Merck
  • PRC1-mediated epigenetic programming is required to generate the ovarian reserve.
PRC1-mediated epigenetic programming is required to generate the ovarian reserve.
Nature communications (2022-08-11)
Mengwen Hu, Yu-Han Yeh, Yasuhisa Munakata, Hironori Abe, Akihiko Sakashita, So Maezawa, Miguel Vidal, Haruhiko Koseki, Neil Hunter, Richard M Schultz, Satoshi H Namekawa
ABSTRACT
The ovarian reserve defines the female reproductive lifespan, which in humans spans decades due to robust maintenance of meiotic arrest in oocytes residing in primordial follicles. Epigenetic reprogramming, including DNA demethylation, accompanies meiotic entry, but the chromatin changes that underpin the generation and preservation of ovarian reserves are poorly defined. We report that the Polycomb Repressive Complex 1 (PRC1) establishes repressive chromatin states in perinatal mouse oocytes that directly suppress the gene expression program of meiotic prophase-I and thereby enable the transition to dictyate arrest. PRC1 dysfuction causes depletion of the ovarian reserve and leads to premature ovarian failure. Our study demonstrates a fundamental role for PRC1-mediated gene silencing in female reproductive lifespan, and reveals a critical window of epigenetic programming required to establish ovarian reserve.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein