Merck
  • The onset of circulation triggers a metabolic switch required for endothelial to hematopoietic transition.
The onset of circulation triggers a metabolic switch required for endothelial to hematopoietic transition.
Cell reports (2021-12-16)
Emanuele Azzoni, Vincent Frontera, Giorgio Anselmi, Christina Rode, Chela James, Elitza M Deltcheva, Atanasiu S Demian, John Brown, Cristiana Barone, Arianna Patelli, Joe R Harman, Matthew Nicholls, Simon J Conway, Edward Morrissey, Sten Eirik W Jacobsen, Duncan B Sparrow, Adrian L Harris, Tariq Enver, Marella F T R de Bruijn
ABSTRACT
Hematopoietic stem cells (HSCs) emerge during development from the vascular wall of the main embryonic arteries. The onset of circulation triggers several processes that provide critical external factors for HSC generation. Nevertheless, it is not fully understood how and when the onset of circulation affects HSC emergence. Here we show that in Ncx1-/- mouse embryos devoid of circulation the HSC lineage develops until the phenotypic pro-HSC stage. However, these cells reside in an abnormal microenvironment, fail to activate the hematopoietic program downstream of Runx1, and are functionally impaired. Single-cell transcriptomics shows that during the endothelial-to-hematopoietic transition, Ncx1-/- cells fail to undergo a glycolysis to oxidative phosphorylation metabolic switch present in wild-type cells. Interestingly, experimental activation of glycolysis results in decreased intraembryonic hematopoiesis. Our results suggest that the onset of circulation triggers metabolic changes that allow HSC generation to proceed.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
DMOG, ≥98% (HPLC)