Merck
  • Vitamin D-Dimer: A Possible Biomolecule Modulator in Cytotoxic and Phagocytosis Processes?
Vitamin D-Dimer: A Possible Biomolecule Modulator in Cytotoxic and Phagocytosis Processes?
Biomedicines (2022-07-28)
Ralf Herwig, Katharina Erlbacher, Amela Ibrahimagic, Mehtap Kacar, Naime Brajshori, Petrit Beqiri, Joachim Greilberger
ABSTRACT
Vitamin D3 complexed to deglycosylated vitamin D binding protein (VitD-dgVDBP) is a water-soluble vitamin D dimeric compound (VitD-dgVDBP). It is not clear how VitD-dgVDBP affects circulating monocytes, macrophages, other immune cell systems, including phagocytosis and apoptosis, and the generation of reactive oxygen species (ROS) compared to dgVDBP. Flow cytometry was used to measure superoxide anion radical (O2*-) levels and macrophage activity in the presence of VitD-dgVDBP or dgVDBP. VitD-dgVDBP was incubated with normal human lymphocytes (nPBMCs), and several clusters of determination (CDs) were estimated. dgVDBP and VitD-dgVDBP apoptosis was estimated on malignant prostatic cells. The macrophage activity was 2.8-fold higher using VitD-dgVDBP (19.8·106 counts) compared to dgVDBP (7.0·106 counts), but O2*- production was 1.8-fold lower in favor of VitD-dgVDBP (355·103 counts) compared to dgVDBP (630·106 counts). The calculated ratio of the radical/macrophage activity was 5-fold lower compared to that of dgVDBP. Only VitD-dgVDBP activated caspase-3 (8%), caspase-9 (13%), and cytochrome-C (11%) on prostatic cancer cells. PE-Cy7-labeled VitD-dgVDBP was found to bind to cytotoxic suppressor cells, monocytes/macrophages, dendritic and natural killer cells (CD8+), and helper cells (CD4+). After 12 h of co-incubation of nPBMCs with VitD-dgVDBP, significant activation and expression were measured for CD16++/CD16 (0.6 ± 0.1% vs. 0.4 ± 0.1%, p < 0.05), CD45k+ (96.0 ± 6.0% vs. 84.7 ± 9.5%, p < 0.05), CD85k+ (24.3 ± 13.2% vs. 3.8 ± 3.2%, p < 0.05), and CD85k+/CD123+ (46.8 ± 8.1% vs. 3.5 ± 3.7%, p < 0.001) compared to the control experiment. No significant difference was found using CD3+, CD4+, CD8+, CD4/CD8, CD4/CD8, CD16+, CD16++, CD14+, or CD123+. A significant decline in CD14+/CD16+ was obtained in the presence of VitD-dgVDBP (0.7 ± 0.2% vs. 3.1 ± 1.7%; p < 0.01). The newly developed water-soluble VitD3 form VitD-dgVDBP affected cytotoxic suppressor cells by activating the low radical-dependent CD16 pathway and seemed to induce apoptosis in malignant prostatic cells.
Sigma-Aldrich
Rabbit Anti-Goat IgG Antibody, HRP conjugate, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-Vitamin D-Binding Protein/GC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Gc-globulin from human plasma, ≥90% (SDS-PAGE), lyophilized powder (containing sodium chloride and sodium phosphate buffer salt)