Merck
  • Thorium inhibits human respiratory chain complex IV (cytochrome c oxidase).
Thorium inhibits human respiratory chain complex IV (cytochrome c oxidase).
Journal of hazardous materials (2021-12-10)
Libing Yu, Zhaozhu Lin, Xuedan Cheng, Jian Chu, Xijian Li, Chun Chen, Tinghua Zhu, Wenjing Li, Wei Lin, Wei Tang
ABSTRACT
Thorium is a radioactive heavy metal and an emerging environmental pollutant. Ecological and human health risks from thorium exposure are growing with the excavation of rare earth metals and implementation of thorium-based nuclear reactors. Thorium poisoning is associated with carcinogenesis, liver impairments, and congenital anomalies. To date, the biomolecular targets that underlie thorium-induced toxicity remain unknown. Here, we used in vitro enzymatic activity assays to comprehensively evaluate the effects of thorium on the mitochondrial respiration process. Thorium was found to inhibit respiratory chain complex IV (cytochrome c oxidase) at sub-micromolar concentrations (IC50 ~ 0.4 μM, 90 μg/L). This is lower than the thorium level limit (246 μg/L) in drinking water specified by the World Health Organization. The inhibitory effects were further verified in mitochondria from human bone and liver cells (thorium mainly deposits in these organs). The inhibition of cytochrome c oxidase can readily rationalize well-documented cellular toxicities of thorium, such as alteration of mitochondrial membrane potential and production of reactive oxygen species. Therefore, cytochrome c oxidase is potentially a key molecular target underlying thorium-induced toxicological effect.
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein