Merck
Epithelial cell shape is regulated by Lulu proteins via myosin-II.
Journal of cell science (2010-01-28)
Hiroyuki Nakajima, Takuji Tanoue
ABSTRACT
Cell-shape change in epithelial structures is fundamental to animal morphogenesis. Recent studies identified myosin-II as the major generator of driving forces for cell-shape changes during morphogenesis. Lulu (Epb41l5) is a major regulator of morphogenesis, although the downstream molecular and cellular mechanisms remain obscure in mammals. In Drosophila and zebrafish, Lulu proteins were reported to negatively regulate Crumbs, an apical domain regulator, thus regulating morphogenesis. In this study, we show that mammalian Lulu activates myosin-II, thus regulating epithelial cell shape. In our experiments, Lulu expression in epithelial cells resulted in apical constriction and lateral elongation in the cells, accompanied by upregulation of myosin-II. The inhibition of myosin-II activity almost completely blocked this Lulu-driven cell-shape change. We further found that Rock participates in the myosin-II activation. Additionally, RNAi-mediated depletion of Lulu in epithelial cells resulted in disorganization of myosin-II and a concomitant loss of proper lateral domain organization in the cells. From these results, we propose that Lulu regulates epithelial cell shape by controlling myosin-II activity.
Sigma-Aldrich
Anti-PAR-3 Antibody, clone 8E8, clone 8E8, from mouse