Merck
  • Single-cell analysis of human skin identifies CD14+ type 3 dendritic cells co-producing IL1B and IL23A in psoriasis.
Single-cell analysis of human skin identifies CD14+ type 3 dendritic cells co-producing IL1B and IL23A in psoriasis.
The Journal of experimental medicine (2021-07-20)
Satoshi Nakamizo, Charles-Antoine Dutertre, Ahad Khalilnezhad, Xiao Meng Zhang, Shawn Lim, Josephine Lum, Geraldine Koh, Charlene Foong, Pearly Jean Ai Yong, Kahbing Jasmine Tan, Reiko Sato, Kaori Tomari, Laurent Yvan-Charvet, Helen He, Emma Guttman-Yassky, Benoit Malleret, Rintaro Shibuya, Masashi Iwata, Baptiste Janela, Tsuyoshi Goto, Tan Siyun Lucinda, Mark B Y Tang, Colin Theng, Valerie Julia, Feriel Hacini-Rachinel, Kenji Kabashima, Florent Ginhoux
ABSTRACT
Inflammatory skin diseases including atopic dermatitis (AD) and psoriasis (PSO) are underpinned by dendritic cell (DC)-mediated T cell responses. Currently, the heterogeneous human cutaneous DC population is incompletely characterized, and its contribution to these diseases remains unclear. Here, we performed index-sorted single-cell flow cytometry and RNA sequencing of lesional and nonlesional AD and PSO skin to identify macrophages and all DC subsets, including the newly described mature LAMP3+BIRC3+ DCs enriched in immunoregulatory molecules (mregDC) and CD14+ DC3. By integrating our indexed data with published skin datasets, we generated a myeloid cell universe of DC and macrophage subsets in healthy and diseased skin. Importantly, we found that CD14+ DC3s increased in PSO lesional skin and co-produced IL1B and IL23A, which are pathological in PSO. Our study comprehensively describes the molecular characteristics of macrophages and DC subsets in AD and PSO at single-cell resolution, and identifies CD14+ DC3s as potential promoters of inflammation in PSO.