Merck
  • Chloroplast Proteome of Nicotiana benthamiana Infected by Tomato Blistering Mosaic Virus.
Chloroplast Proteome of Nicotiana benthamiana Infected by Tomato Blistering Mosaic Virus.
The protein journal (2018-05-29)
Esau Megias, Lílian Silveira Travassos do Carmo, Cícero Nicolini, Luciano Paulino Silva, Rosana Blawid, Tatsuya Nagata, Angela Mehta
ABSTRACT
Tymovirus is a genus of plant pathogenic viruses that infects several dicotyledonous plants worldwide, causing serious diseases in economically important crops. The known cytopathic effect on the host cell organelles involves chloroplast membrane deformation and the induction of vesicles in its periphery. These vesicles are known to be the location where tymoviral genomic RNA replication occurs. Tomato blistering mosaic virus (ToBMV) is a tymovirus recently identified in tomato plants in Brazil, which is able to infect several other plants, including tobacco. In this work, we investigated the chloroplast proteomic profile of ToBMV-infected N. benthamiana using bidimensional electrophoresis (2-DE) and mass spectrometry, aiming to study the virus-host interaction related to the virus replication and infection. A total of approximately 200 spots were resolved, out of which 36 were differentially abundant. Differential spots were identified by mass spectrometry including photosynthesis-related and defense proteins. We identified proteins that may be targets of a direct interaction with viral proteins, such as ATP synthase β subunit, RNA polymerase beta-subunit, 50S ribosomal protein L6 and Trigger factor-like protein. The identification of these candidate proteins gives support for future protein-protein interaction studies to confirm their roles in virus replication and disease development.
Sigma-Aldrich
Chloroplast Isolation Kit, isolation of intact chloroplasts from leaves