Merck
  • Ligand-mediated PAI-1 inhibition in a mouse model of peritoneal carcinomatosis.
Ligand-mediated PAI-1 inhibition in a mouse model of peritoneal carcinomatosis.
Cell reports. Medicine (2022-03-05)
Josephine Hendrikson, Ying Liu, Wai Har Ng, Jing Yi Lee, Abner Herbert Lim, Jui Wan Loh, Cedric C Y Ng, Whee Sze Ong, Joey Wee-Shan Tan, Qiu Xuan Tan, Gillian Ng, Nicholas B Shannon, Weng Khong Lim, Tony K H Lim, Clarinda Chua, Jolene Si Min Wong, Grace Hwei Ching Tan, Jimmy Bok Yan So, Khay Guan Yeoh, Bin Tean Teh, Claramae Shulyn Chia, Khee Chee Soo, Oi Lian Kon, Iain Beehuat Tan, Jason Yongsheng Chan, Melissa Ching Ching Teo, Chin-Ann J Ong
ABSTRACT
Peritoneal carcinomatosis (PC) present a ubiquitous clinical conundrum in all intra-abdominal malignancies. Via functional and transcriptomic experiments of ascites-treated PC cells, we identify STAT3 as a key signaling pathway. Integrative analysis of publicly available databases and correlation with clinical cohorts (n = 7,359) reveal putative clinically significant activating ligands of STAT3 signaling. We further validate a 3-biomarker prognostic panel in ascites independent of clinical covariates in a prospective study (n = 149). Via single-cell sequencing experiments, we uncover that PAI-1, a key component of the prognostic biomarker panel, is largely secreted by fibroblasts and mesothelial cells. Molecular stratification of ascites using PAI-1 levels and STAT3 activation in ascites-treated cells highlight a therapeutic opportunity based on a phenomenon of paracrine addiction. These results are recapitulated in patient-derived ascites-dependent xenografts. Here, we demonstrate therapeutic proof of concept of direct ligand inhibition of a prognostic target within an enclosed biological space.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture