Merck
  • Determination of seven pyrethroids biocides and their synergist in indoor air by thermal-desorption gas chromatography/mass spectrometry after sampling on Tenax TA ® passive tubes.
Determination of seven pyrethroids biocides and their synergist in indoor air by thermal-desorption gas chromatography/mass spectrometry after sampling on Tenax TA ® passive tubes.
Talanta (2014-10-05)
Caroline Raeppel, Brice M Appenzeller, Maurice Millet
ABSTRACT
A method coupling thermal desorption and gas chromatography/mass spectrometry (GC/MS) was developed for the simultaneous determination of 7 pyrethroids (allethrin, bifenthrin, cyphenothrin, imiprothrin, permethrin, prallethrin and tetramethrin) and piperonyl butoxide adsorbed on Tenax TA(®) passive samplers after exposure in indoor air. Thermal desorption was selected as it permits efficient and rapid extraction without solvent used together with a good sensitivity. Detection (S/N>3) and quantification (S/N>10) limits varied between 0.001 ng and 2.5 ng and between 0.005 and 10 ng respectively with a reproducibility varied between 14% (bifenthrin) and 39% (permethrin). The method was used for the comparison indoor air contamination after low-pressure spraying and fumigation application in a rubbish chute situated in the basement of a building.
Product Description
Supelco
Cyphenothrin, PESTANAL®, analytical standard, sum of isomers