Merck
  • Protective role of arzanol against lipid peroxidation in biological systems.
Protective role of arzanol against lipid peroxidation in biological systems.
Chemistry and physics of lipids (2010-10-12)
Antonella Rosa, Federica Pollastro, Angela Atzeri, Giovanni Appendino, M Paola Melis, Monica Deiana, Alessandra Incani, Debora Loru, M Assunta Dessì
ABSTRACT
This study examines the protective effect of arzanol, a pyrone-phloroglucinol etherodimer from Helichrysum italicum subsp. microphyllum, against the oxidative modification of lipid components induced by Cu(2+) ions in human low density lipoprotein (LDL) and by tert-butyl hydroperoxide (TBH) in cell membranes. LDL pre-treatment with arzanol significantly preserved lipoproteins from oxidative damage at 2h of oxidation, and showed a remarkable protective effect on the reduction of polyunsaturated fatty acids and cholesterol levels, inhibiting the increase of oxidative products (conjugated dienes fatty acids hydroperoxides, 7β-hydroxycholesterol, and 7-ketocholesterol). Arzanol, at non-cytotoxic concentrations, exerted a noteworthy protection on TBH-induced oxidative damage in a line of fibroblasts derived from monkey kidney (Vero cells) and in human intestinal epithelial cells (Caco-2), decreasing, in both cell lines, the formation of oxidative products (hydroperoxides and 7-ketocholesterol) from the degradation of unsaturated fatty acids and cholesterol. The cellular uptake and transepithelial transport of the compound were also investigated in Caco-2 cell monolayers. Arzanol appeared to accumulate in Caco-2 epithelial cells. This phenol was able to pass through the intestinal Caco-2 monolayers, the apparent permeability coefficients (P(app)) in the apical-to-basolateral and basolateral-to-apical direction at 2h were 1.93±0.36×10(-5) and 2.20±0.004×10(-5)cm/s, respectively, suggesting a passive diffusion pathway. The results of the work qualify arzanol as a potent natural antioxidant with a protective effect against lipid oxidation in biological systems.
Sigma-Aldrich
Arzanol, from Helichrysum italicum, ≥98%