- Method development for the determination of selected pesticides on tobacco by high-performance liquid chromatography-electrospray ionisation-tandem mass spectrometry.
Method development for the determination of selected pesticides on tobacco by high-performance liquid chromatography-electrospray ionisation-tandem mass spectrometry.
A method was developed for the quantitative determination of alachlor, benalaxyl, clomazone, diflubenzuron, dimethomorph, diphenamid, ethofumesate, metalaxyl, methoprene, metobromuron and piperonyl butoxide on tobacco. The pesticides were extracted with water and methanol from five different types of tobacco. The extracts were purified by partition on an extraction cartridge containing diatomaceous earth. The purified extracts were analysed by reversed-phase high-performance liquid chromatography connected to an atmospheric pressure ionisation-electrospray-triple quadrupole mass spectrometer operating in the positive ion mode. Two different transitions and their relative intensities were monitored for unambiguous identification. All pesticides presented overall recovery rates between 35% and 110%. The trueness is near 100% and the interday precision is below 15%. The limits of quantifications are equal or below the guidance residue levels proposed by the Agrochemical Advisory Committee of CORESTA, an association of organisations having scientific research relative to tobacco.