Merck
  • Mechanistic Studies of the Multiple Myeloma and Melanoma Cell-Selective Toxicity of the Rpn13-Binding Peptoid KDT-11.
Mechanistic Studies of the Multiple Myeloma and Melanoma Cell-Selective Toxicity of the Rpn13-Binding Peptoid KDT-11.
Cell chemical biology (2020-08-29)
Paige Dickson, Scott Simanski, John Maina Ngundu, Thomas Kodadek
ABSTRACT
We previously reported a peptoid ligand for the proteasomal ubiquitin receptor Rpn13 called KDT-11 and demonstrated that this compound is toxic to multiple myeloma cells, but not non-malignant cells. Here, we show that KDT-11 decreases the viability of a variety of cancer cell lines, especially melanomas and various blood cancers. The peptoid induces selective G1 cell-cycle arrest, resulting in eventual apoptosis. While KDT-11 does not antagonize any of the known protein-protein interactions involving Rpn13, the peptoid inhibits the ability of Rpn13 to stimulate the activity of an associated deubiquitylase Uch37/UCHL5 in vitro, suggesting a high level of Uch37 activity might be important for cancer cell proliferation. However, a variety of experiments in SK-MEL-5 melanoma cells suggest that KDT-11's cytotoxic effects are mediated by interactions with proteins other than Rpn13.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
KOD Hot Start Master Mix, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., ready-to-use solution, suitable for PCR
Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder