Merck
  • Molecular detection of Treponema denticola and Porphyromonas gingivalis in carotid and aortic atheromatous plaques by FISH: report of two cases.
Molecular detection of Treponema denticola and Porphyromonas gingivalis in carotid and aortic atheromatous plaques by FISH: report of two cases.
Journal of medical microbiology (2004-12-14)
Francesca Cavrini, Vittorio Sambri, Annette Moter, Dora Servidio, Antonella Marangoni, Lucio Montebugnoli, Federico Foschi, Carlo Prati, Roberto Di Bartolomeo, Roberto Cevenini
ABSTRACT
Treponema denticola and Porphyromonas gingivalis have been identified in atheromatous plaques of two patients suffering from atherosclerosis by PCR and fluorescence in situ hybridization (FISH). The use of the FISH technique suggested that these periodontopathic micro-organisms might be metabolically active within the wall of arteries, under the atherosclerotic lesion.
Sigma-Aldrich
Eubacteria FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH), 20 µM in water
Sigma-Aldrich
Porphyromonas gingivalis FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Porphyromonas gingivalis FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH)