Merck
  • Fitness analyses of all possible point mutations for regions of genes in yeast.
Fitness analyses of all possible point mutations for regions of genes in yeast.
Nature protocols (2012-06-23)
Ryan Hietpas, Benjamin Roscoe, Li Jiang, Daniel N A Bolon
ABSTRACT
Deep sequencing can accurately measure the relative abundance of hundreds of mutations in a single bulk competition experiment, which can give a direct readout of the fitness of each mutant. Here we describe a protocol that we previously developed and optimized to measure the fitness effects of all possible individual codon substitutions for 10-aa regions of essential genes in yeast. Starting with a conditional strain (i.e., a temperature-sensitive strain), we describe how to efficiently generate plasmid libraries of point mutants that can then be transformed to generate libraries of yeast. The yeast libraries are competed under conditions that select for mutant function. Deep-sequencing analyses are used to determine the relative fitness of all mutants. This approach is faster and cheaper per mutant compared with analyzing individually isolated mutants. The protocol can be performed in ∼4 weeks and many 10-aa regions can be analyzed in parallel.
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture