Merck
  • Bombesin receptors in distinct tissue compartments of human pancreatic diseases.
Bombesin receptors in distinct tissue compartments of human pancreatic diseases.
Laboratory investigation; a journal of technical methods and pathology (2001-01-05)
A Fleischmann, U Läderach, H Friess, M W Buechler, J C Reubi
ABSTRACT
Overexpression of receptors for regulatory peptides in various human diseases is reportedly of clinical interest. Among these peptides, bombesin and gastrin-releasing peptide (GRP) have been shown to play a physiological and pathophysiological role in pancreatic tissues. Our aim has been to localize bombesin receptors in the human diseased pancreas to identify potential clinical applications of bombesin analogs in this tissue. The presence of bombesin receptor subtypes has been evaluated in specimens of human pancreatic tissues with chronic pancreatitis (n = 23) and ductal pancreatic carcinoma (n = 29) with in vitro receptor autoradiography on tissue sections incubated with 125I-[Tyr4]-bombesin or the universal ligand 125I-[D-Tyr6, beta-Ala11, Phe13, Nle14]-bombesin(6-14) as radioligands and displaced by subtype-selective bombesin receptor agonists and antagonists. GRP receptors were identified in the pancreatic exocrine parenchyma in 17 of 20 cases with chronic pancreatitis. No measurable bombesin receptors were found in the tumor tissue of ductal pancreatic carcinomas, however, GRP receptors were detected in a subset of peritumoral small veins in 19 of 29 samples. Moreover, residual pancreatic islets in these tissues were shown to express the BB3 receptor subtype. These data demonstrate the presence of bombesin receptors in three distinct tissue compartments of the pancreas, namely GRP receptors in the exocrine parenchyma in chronic pancreatitis and in peritumoral vessels around ductal pancreatic carcinomas, and BB3 receptors in residual pancreatic islets. Such a selective expression of bombesin receptor subtypes in pancreatic tissues may not only be of pathophysiological significance but may represent the basis for potential diagnostic and therapeutic clinical applications of bombesin analogs, including GRP receptor scintigraphy to differentiate chronic pancreatitis from ductal pancreatic carcinoma.
Sigma-Aldrich
[Tyr4]-Bombesin, ≥97% (HPLC)