Merck
  • Lipoteichoic Acid Isolated from Lactobacillus plantarum Maintains Inflammatory Homeostasis through Regulation of Th1- and Th2-Induced Cytokines.
Lipoteichoic Acid Isolated from Lactobacillus plantarum Maintains Inflammatory Homeostasis through Regulation of Th1- and Th2-Induced Cytokines.
Journal of microbiology and biotechnology (2018-11-13)
Ji Eun Ahn, Hangeun Kim, Dae Kyun Chung
ABSTRACT
Lipoteichoic acid isolated from Lactobacillus plantarum K8 (pLTA) alleviates lipopolysaccharide (LPS)-induced excessive inflammation through inhibition of TNF-α and interleukin (IL)-6. In addition, pLTA increases the survival rate of mice in a septic shock model. In the current study, we have found that pLTA contributes to homeostasis through regulation of pro- and anti-inflammatory cytokine production. In detail, pLTA decreased the production of IL-10 by phorbol-12-myristate-13-acetate (PMA)-differentiated THP-1 cells stimulated with prostaglandin E2 (PGE-2) and LPS. However, TNF-α production which was inhibited by PGE-2+LPS increased by pLTA treatment. The regulatory effects of IL-10 and TNF-α induced by PGE-2 and LPS in PMA-differentiated THP-1 cells were mediated by pLTA, but not by other LTAs isolated from either Staphylococcus aureus (aLTA) or L. sakei (sLTA). Further studies revealed that pLTA-mediated IL-10 inhibition and TNF-α induction in PGE-2+LPS-stimulated PMA-differentiated THP-1 cells were mediated by dephosphorylation of p38 and phosphorylation of c-Jun N-terminal kinase (JNK), respectively. Reduction of pLTA-mediated IL-10 inhibited the metastasis of breast cancer cells (MDA-MB-231), which was induced by IL-10 or conditioned media prepared from PGE-2+LPS-stimulated PMA-differentiated THP-1 cells. Taken together, our data suggest that pLTA contributes to inflammatory homeostasis through induction of repressed pro-inflammatory cytokines as well as inhibition of excessive anti-inflammatory cytokines.
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).