Merck
  • Glioblastomas acquire myeloid-affiliated transcriptional programs via epigenetic immunoediting to elicit immune evasion.
Glioblastomas acquire myeloid-affiliated transcriptional programs via epigenetic immunoediting to elicit immune evasion.
Cell (2021-04-16)
Ester Gangoso, Benjamin Southgate, Leanne Bradley, Stefanie Rus, Felipe Galvez-Cancino, Niamh McGivern, Esra Güç, Chantriolnt-Andreas Kapourani, Adam Byron, Kirsty M Ferguson, Neza Alfazema, Gillian Morrison, Vivien Grant, Carla Blin, IengFong Sou, Maria Angeles Marques-Torrejon, Lucia Conde, Simona Parrinello, Javier Herrero, Stephan Beck, Sebastian Brandner, Paul M Brennan, Paul Bertone, Jeffrey W Pollard, Sergio A Quezada, Duncan Sproul, Margaret C Frame, Alan Serrels, Steven M Pollard
ABSTRACT
Glioblastoma multiforme (GBM) is an aggressive brain tumor for which current immunotherapy approaches have been unsuccessful. Here, we explore the mechanisms underlying immune evasion in GBM. By serially transplanting GBM stem cells (GSCs) into immunocompetent hosts, we uncover an acquired capability of GSCs to escape immune clearance by establishing an enhanced immunosuppressive tumor microenvironment. Mechanistically, this is not elicited via genetic selection of tumor subclones, but through an epigenetic immunoediting process wherein stable transcriptional and epigenetic changes in GSCs are enforced following immune attack. These changes launch a myeloid-affiliated transcriptional program, which leads to increased recruitment of tumor-associated macrophages. Furthermore, we identify similar epigenetic and transcriptional signatures in human mesenchymal subtype GSCs. We conclude that epigenetic immunoediting may drive an acquired immune evasion program in the most aggressive mesenchymal GBM subtype by reshaping the tumor immune microenvironment.
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Roche
Collagenase D, from Clostridium histolyticum
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Goat Serum Donor Herd, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%