Merck
  • Paired heavy- and light-chain signatures contribute to potent SARS-CoV-2 neutralization in public antibody responses.
Paired heavy- and light-chain signatures contribute to potent SARS-CoV-2 neutralization in public antibody responses.
Cell reports (2021-09-30)
Bailey B Banach, Gabriele Cerutti, Ahmed S Fahad, Chen-Hsiang Shen, Matheus Oliveira De Souza, Phinikoula S Katsamba, Yaroslav Tsybovsky, Pengfei Wang, Manoj S Nair, Yaoxing Huang, Irene M Francino-Urdániz, Paul J Steiner, Matías Gutiérrez-González, Lihong Liu, Sheila N López Acevedo, Alexandra F Nazzari, Jacy R Wolfe, Yang Luo, Adam S Olia, I-Ting Teng, Jian Yu, Tongqing Zhou, Eswar R Reddem, Jude Bimela, Xiaoli Pan, Bharat Madan, Amy D Laflin, Rajani Nimrania, Kwok-Yung Yuen, Timothy A Whitehead, David D Ho, Peter D Kwong, Lawrence Shapiro, Brandon J DeKosky
ABSTRACT
Understanding mechanisms of protective antibody recognition can inform vaccine and therapeutic strategies against SARS-CoV-2. We report a monoclonal antibody, 910-30, targeting the SARS-CoV-2 receptor-binding site for ACE2 as a member of a public antibody response encoded by IGHV3-53/IGHV3-66 genes. Sequence and structural analyses of 910-30 and related antibodies explore how class recognition features correlate with SARS-CoV-2 neutralization. Cryo-EM structures of 910-30 bound to the SARS-CoV-2 spike trimer reveal binding interactions and its ability to disassemble spike. Despite heavy-chain sequence similarity, biophysical analyses of IGHV3-53/3-66-encoded antibodies highlight the importance of native heavy:light pairings for ACE2-binding competition and SARS-CoV-2 neutralization. We develop paired heavy:light class sequence signatures and determine antibody precursor prevalence to be ∼1 in 44,000 human B cells, consistent with public antibody identification in several convalescent COVID-19 patients. These class signatures reveal genetic, structural, and functional immune features that are helpful in accelerating antibody-based medical interventions for SARS-CoV-2.
Sigma-Aldrich
Monoclonal Anti-MYC-FITC , (C-terminal) antibody produced in mouse, clone 9E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-FITC antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous solution