Merck
  • An alcoholic extract of Thuja orientalis L. leaves inhibits autophagy by specifically targeting pro-autophagy PIK3C3/VPS34 complex.
An alcoholic extract of Thuja orientalis L. leaves inhibits autophagy by specifically targeting pro-autophagy PIK3C3/VPS34 complex.
Scientific reports (2021-09-08)
Juneyoung Jung, Yoomi Chun, Young Pyo Jang, Myung Sook Oh, Jeong Hee Kim, Joungmok Kim
ABSTRACT
Autophagy is a lysosome-dependent degradation program to maintain cellular homeostasis in response to a variety of stressful conditions, such as long-lived or non-functional subcellular organelles, protein aggregates, nutrient limitation, and virus/bacteria infection. Accordingly, dysregulation of autophagy is closely associated with many human pathophysiological conditions, such as neurodegenerative diseases, aging, and cancer, and autophagy is highlighted as an important therapeutic target for these human diseases. In autophagy process, PIK3C3/VPS34 complex plays important roles in autophagosome biogenesis. Accumulating evidences that inhibition of PIK3C3/VPS34 complex successfully blocks autophagy make the complex as an attractive target for the development of autophagy-specific inhibitors. However, considering that various forms of PIK3C3/VPS34 complex exist and they are involved in many different cellular functions, the targeting of the pro-autophagy PIK3C3/VPS34 complex is required to specifically inhibit autophagy. To identify autophagy inhibitors targeting the pro-autophagy complex, we have performed the screening of a customized natural product library consisting of 35 herbal extracts which are widely used in the oriental medicine as anti-inflammation and/or anti-tumor reagents. We discovered that an alcoholic extract of Thuja orientalis L. leaves inhibits pro-autophagy complex formation by disrupting the interaction between autophagy-specific factor, ATG14L, and the complex core unit Vps34-Beclin 1 in vitro. Also, it inhibits the nutrient starvation induced autophagy and diminished pro-autophagy PIK3C3/VPS34 complex containing either ATG14L or UVRAG in several cell lines. Our results strongly suggest that Thuja orientalis L. leave extract functions as an autophagy-specific inhibitor not decreasing the complex activity nor the protein level, but preventing protein-protein interaction between autophagy-specific factor (ATG14L and UVRAG) and PIK3C3/VPS34 complex core unit, Vps34-Beclin 1, thereby specifically depleting the pro-autophagy complex to inhibit autophagy.
Sigma-Aldrich
3-Carene, 90%
Sigma-Aldrich
(−)-trans-Caryophyllene, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution