Merck
  • Interleukin-8 Predicts Fatigue at 12 Months Post-Injury in Children with Traumatic Brain Injury.
Interleukin-8 Predicts Fatigue at 12 Months Post-Injury in Children with Traumatic Brain Injury.
Journal of neurotrauma (2019-12-28)
Alison Crichton, Vera Ignjatovic, Franz E Babl, Ed Oakley, Mardee Greenham, Stephen Hearps, Carmel Delzoppo, Miriam H Beauchamp, Anne-Marie Guerguerian, Kathy Boutis, Evyatar Hubara, Jamie Hutchison, Vicki Anderson
ABSTRACT
Despite many children experiencing fatigue after childhood brain injury, little is known about the predictors of this complaint. To date, traditional indices of traumatic brain injury (TBI) severity have not predicted reliably persisting fatigue (up to three years post-injury). This study aimed to establish whether persisting fatigue is predicted by serum biomarker concentrations in child TBI. We examined whether acute serum biomarker expression would improve prediction models of 12-month fatigue based on injury severity. Blood samples were collected from 87 children (1-17 years at injury) sustaining mild to severe TBI (Glasgow Coma Scale [GCS] range 3-15; mean 12.43; classified as mild TBI [n = 50, 57%] vs. moderate/severe TBI [n = 37, 43%]), and presenting to the emergency departments (ED) and pediatric intensive care units (PICU) at one of three tertiary pediatric hospitals (Royal Children's Hospital (RCH); Hospital for Sick Children (HSC), Toronto; St Justine Children's Hospital (SJH), Montreal). Six serum biomarker concentrations were measured within 24 h of injury (interleukin-6, interleukin-8 [IL-8], soluble vascular cell adhesion molecule [SVCAM], S100 calcium binding protein B [S100B], neuron specific enolase [NSE], and soluble neural cell adhesion molecule [sNCAM]). Fatigue at 12 months post-injury was measured using the Pediatric Quality of Life Inventory Multidimensional Fatigue Scale (parent report), classified as present/absent using previously derived cut-points. At 12 months post-injury, 22% of participants experienced fatigue. A model including IL-8 was the best serum biomarker for estimating the probability of children experiencing fatigue at 12 months post-injury. The IL-8 also significantly improved predictive models of fatigue based on severity.
Product Description
Millipore
MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 3 - Neuroscience Multiplex Assay, The analytes available for this multiplex kit are: BDNF, Cathepsin D, MPO, NCAM, PAI-1 (total), PDGF-AA, PDGF-AB/BB, RANTES, sICAM-1, sVCAM-1.
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Millipore
Human S100B ELISA, EZHS100B-33K, This Human S100B ELISA kit is to be used for the quantification of human S100B in cerebrospinal fluid, serum & plasma.