- Implantation of Human Peripheral Corneal Spheres into Cadaveric Human Corneal Tissue.
Implantation of Human Peripheral Corneal Spheres into Cadaveric Human Corneal Tissue.
Bio-protocol (2017-07-20)
Jeremy John Mathan, Salim Ismail, Jennifer Jane McGhee, Trevor Sherwin
PMID34541142
ABSTRACT
Stem and progenitor cells isolated from human limbal tissue can be cultured in vitro as spheres. These spheres have potential for use as transplantable elements for the repopulation of corneal tissue ( Mathan et al., 2016 ). Herein we describe the detailed protocol for the implantation of human corneal spheres into cadaveric human corneal tissue. This protocol describes the procedure for sphere formation and culture, preparation of tissue for sphere implantation, corneal limbus microsurgery and sphere implantation.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid