Merck
  • Guanidinoneomycin-maleimide molecular transporter: synthesis, chemistry and cellular uptake.
Guanidinoneomycin-maleimide molecular transporter: synthesis, chemistry and cellular uptake.
Organic & biomolecular chemistry (2021-07-14)
Kaivin Hadidi, Maria Cristina Bellucci, Sergio Dall'Angelo, Alasdair Leeson-Payne, Justin J Rochford, Jeffery D Esko, Yitzhak Tor, Alessandro Volonterio
ABSTRACT
Guanidinoglycosides are a class of non-cytotoxic molecular transporters capable of delivering high molecular weight bioactive cargos into cells at low nanomolar concentrations. Efficient bioconjugation with guanidinoglycosides has been previously demonstrated by utilizing a guanidinoneomycin decorated with a reactive but also unstable N-hydroxysuccinimmide ester-containing linker. Herein we report the synthesis, chemistry, and application of a new, stable guanidinoneomycin derivative armed with a highly specific maleimide moiety which allows for thiol-maleimide click chemistry, a highly popular bioconjugation strategy, widening the field of application of these intriguing and useful delivery vehicles.
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture