Merck
  • High membrane expression of CMTM6 in hepatocellular carcinoma is associated with tumor recurrence.
High membrane expression of CMTM6 in hepatocellular carcinoma is associated with tumor recurrence.
Cancer science (2021-06-04)
Ryo Muranushi, Kenichiro Araki, Takehiko Yokobori, Batbayar Chingunjav, Kouki Hoshino, Gantumur Dolgormaa, Kei Hagiwara, Takahiro Yamanaka, Norihiro Ishii, Mariko Tsukagoshi, Takamichi Igarashi, Akira Watanabe, Norio Kubo, Norifumi Harimoto, Yuki Shimoda, Rie Sano, Tetsunari Oyama, Hiroshi Saeki, Ken Shirabe
ABSTRACT
CKLF-like MARVEL transmembrane domain-containing protein 6 (CMTM6) maintains membrane PD-L1 expression by controlling its endosomal recycling. However, in patients with hepatocellular carcinoma (HCC), the correlation among CMTM6, B7 family ligands, and CD8-positive cytotoxic T lymphocytes (CTLs), and the molecular function of CMTM6 in HCC have not been established. We performed immunohistochemistry to evaluate the relationships among CMTM6 expression, clinicopathological factors, B7 family ligands expression, and CTL infiltration in HCC samples. Moreover, we established CMTM6-knockout human HCC cell lines to evaluate the function of human CMTM6 in immune regulation and tumor viability. CMTM6 expression was positively associated with membrane B7 family ligands expression and CTL infiltration in HCC samples. High CMTM6 expression in HCC tissues was associated with the expression of the proliferation marker Ki-67 and shorter recurrence-free survival. In vitro analysis showed the downregulation of membrane B7 family ligands and proliferation potency in the CMTM6-knockout human HCC cell line. High membrane CMTM6 expression was associated with tumor recurrence and proliferation via the regulation of membranous B7 family ligands expression. Thus, CMTM6 might be a biomarker to predict the risk of HCC recurrence and a therapeutic target to suppress tumor growth and increase CTL activity.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid