Merck
  • Characterization of NADPH Oxidase Expression and Activity in Acute Myeloid Leukemia Cell Lines: A Correlation with the Differentiation Status.
Characterization of NADPH Oxidase Expression and Activity in Acute Myeloid Leukemia Cell Lines: A Correlation with the Differentiation Status.
Antioxidants (Basel, Switzerland) (2021-04-04)
Hassan Dakik, Maya El Dor, Joan Leclerc, Farah Kouzi, Ali Nehme, Margaux Deynoux, Christelle Debeissat, Georges Khamis, Elfi Ducrocq, Aida Ibrik, Marie-José Stasia, Houssam Raad, Hamid Reza Rezvani, Fabrice Gouilleux, Kazem Zibara, Olivier Herault, Frédéric Mazurier
ABSTRACT
In acute myeloid leukemia (AML), a low level of reactive oxygen species (ROS) is associated with leukemic stem cell (LSC) quiescence, whereas a high level promotes blast proliferation. ROS homeostasis relies on a tightly-regulated balance between the antioxidant and oxidant systems. Among the oxidants, NADPH oxidases (NOX) generate ROS as a physiological function. Although it has been reported in AML initiation and development, the contribution of NOX to the ROS production in AML remains to be clarified. The aim of this study was to investigate the NOX expression and function in AML, and to examine the role of NOX in blast proliferation and differentiation. First, we interrogated the NOX expression in primary cells from public datasets, and investigated their association with prognostic markers. Next, we explored the NOX expression and activity in AML cell lines, and studied the impact of NOX knockdown on cell proliferation and differentiation. We found that NOX2 is ubiquitously expressed in AML blasts, and particularly in cells from the myelomonocytic (M4) and monocytic (M5) stages; however, it is less expressed in LSCs and in relapsed AML. This is consistent with an increased expression throughout normal hematopoietic differentiation, and is reflected in AML cell lines. Nevertheless, no endogenous NOX activity could be detected in the absence of PMA stimulation. Furthermore, CYBB knockdown, although hampering induced NOX2 activity, did not affect the proliferation and differentiation of THP-1 and HL-60 cells. In summary, our data suggest that NOX2 is a marker of AML blast differentiation, while AML cell lines lack any NOX2 endogenous activity.
Sigma-Aldrich
Anti-p67-phox Antibody, serum, Upstate®
Sigma-Aldrich
Anti-p47-phox Antibody, serum, Upstate®
Sigma-Aldrich
Anti-p40-phox Antibody, Upstate®, from rabbit