Merck
  • Silencing long non-coding RNA DLX6-AS1 or restoring microRNA-193b-3p enhances thyroid carcinoma cell autophagy and apoptosis via depressing HOXA1.
Silencing long non-coding RNA DLX6-AS1 or restoring microRNA-193b-3p enhances thyroid carcinoma cell autophagy and apoptosis via depressing HOXA1.
Journal of cellular and molecular medicine (2021-09-14)
Ling Feng, Ru Wang, Yifan Wang, Xixi Shen, Qian Shi, Meng Lian, Hongzhi Ma, Jugao Fang
ABSTRACT
Long non-coding RNA DLX6 antisense RNA 1 (DLX6-AS1) lists a critical position in thyroid carcinoma (TC) development. However, the overall comprehension about DLX6-AS1, microRNA (miR)-193b-3p and homeobox A1 (HOXA1) in TC is not thoroughly enough. Concerning to this, this work is pivoted on DLX6-AS1/miR-193b-3p/HOXA1 axis in TC cell growth and autophagy. TC tissues and adjacent normal thyroid tissues were collected, in which expression of DLX6-AS1, miR-193b-3p and HOXA1 was tested, together with their interactions. TC cells were transfected with DLX6-AS1/miR-193b-3p-related oligonucleotides or plasmids to test cell growth and autophagy. Tumorigenesis in nude mice was observed. DLX6-AS1 and HOXA1 were up-regulated, and miR-193b-3p was down-regulated in TC. Depleted DLX6-AS1 or restored miR-193b-3p disturbed cell growth and promoted autophagy. DLX6-AS1 targeted miR-193b-3p and positively regulated HOXA1. miR-193b-3p inhibition mitigated the impaired tumorigenesis induced by down-regulated DLX6-AS1. Tumorigenesis in nude mice was consistent with that in cells. It is clear that DLX6-AS1 depletion hinders TC cell growth and promotes autophagy via up-regulating miR-193b-3p and down-regulating HOXA1.