Merck
  • ATM-phosphorylated SPOP contributes to 53BP1 exclusion from chromatin during DNA replication.
ATM-phosphorylated SPOP contributes to 53BP1 exclusion from chromatin during DNA replication.
Science advances (2021-06-20)
Dejie Wang, Jian Ma, Maria Victoria Botuyan, Gaofeng Cui, Yuqian Yan, Donglin Ding, Yingke Zhou, Eugene W Krueger, Jiang Pei, Xiaosheng Wu, Liguo Wang, Huadong Pei, Mark A McNiven, Dingwei Ye, Georges Mer, Haojie Huang
ABSTRACT
53BP1 activates nonhomologous end joining (NHEJ) and inhibits homologous recombination (HR) repair of DNA double-strand breaks (DSBs). Dissociation of 53BP1 from DSBs and consequent activation of HR, a less error-prone pathway than NHEJ, helps maintain genome integrity during DNA replication; however, the underlying mechanisms are not fully understood. Here, we demonstrate that E3 ubiquitin ligase SPOP promotes HR during S phase of the cell cycle by excluding 53BP1 from DSBs. In response to DNA damage, ATM kinase-catalyzed phosphorylation of SPOP causes a conformational change in SPOP, revealed by x-ray crystal structures, that stabilizes its interaction with 53BP1. 53BP1-bound SPOP induces polyubiquitination of 53BP1, eliciting 53BP1 extraction from chromatin by a valosin-containing protein/p97 segregase complex. Our work shows that SPOP facilitates HR repair over NHEJ during DNA replication by contributing to 53BP1 removal from chromatin. Cancer-derived SPOP mutations block SPOP interaction with 53BP1, inducing HR defects and chromosomal instability.