Merck
  • Novel indoleamine 2,3-dioxygenase-1 inhibitors from a multistep in silico screen.
Novel indoleamine 2,3-dioxygenase-1 inhibitors from a multistep in silico screen.
Bioorganic & medicinal chemistry (2011-11-25)
Jason R Smith, Krystal J Evans, Adam Wright, Robert D Willows, Joanne F Jamie, Renate Griffith
ABSTRACT
Indoleamine 2,3-dioxygenase-1 (IDO-1) is a heme containing enzyme that catalyses the initial step in the major pathway of l-tryptophan catabolism; the kynurenine pathway. A large body of evidence has been accumulating for its immunosuppressive and tumoural escape roles and its applicability as a therapeutic target. Of particular interest is the possibility that IDO-1 inhibition may arrest, and sometimes revert, tumour growth. There exists a continuing need for the development of new and specific inhibitors for IDO-1, and we have created three pharmacophores designed to aid in this search. Initial database hits were further screened using Kier flexibility and a 'What-If' docking technique, designed to overcome the inherent limitations of today's forcefields with regards to heme chemistry. Eighteen compounds were tested in vitro, yielding four novel inhibitors with low micromolar IC(50) values, comparable with current inhibitors.
Sigma-Aldrich
3-Hydroxy-2-naphthoic acid, 98%