- Generation of a human iPSC line, INMi002-A, carrying the most prevalent USH2A variant associated with Usher syndrome type 2.
Generation of a human iPSC line, INMi002-A, carrying the most prevalent USH2A variant associated with Usher syndrome type 2.
Stem cell research (2018-11-24)
Carla Sanjurjo-Soriano, Nejla Erkilic, Gaël Manes, Gregor Dubois, Christian P Hamel, Isabelle Meunier, Vasiliki Kalatzis
PMID30468996
ABSTRACT
We generated an induced pluripotent stem cell (iPSC) line using dermal fibroblasts from a patient with Usher syndrome type 2 (USH2). This individual was homozygous for the most prevalent variant reported in the USH2A gene, c.2299delG localized in exon 13. Reprogramming was performed using the non-integrative Sendai virus reprogramming method and the human OSKM transcription factor cocktail under feeder-free culture conditions. This iPSC line will be an invaluable tool for studying the pathophysiology of USH2 and for testing the efficacy of novel treatments.
Sigma-Aldrich
Monoclonal Anti-AFP antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution