Merck
  • Evaluation of NTF1836 as an inhibitor of the mycothiol biosynthetic enzyme MshC in growing and non-replicating Mycobacterium tuberculosis.
Evaluation of NTF1836 as an inhibitor of the mycothiol biosynthetic enzyme MshC in growing and non-replicating Mycobacterium tuberculosis.
Bioorganic & medicinal chemistry (2011-06-15)
Gerald L Newton, Nancy Buchmeier, James J La Clair, Robert C Fahey
ABSTRACT
The mycothiol biosynthesis enzyme MshC catalyzes the ligation of cysteine with the pseudodisaccharide GlcN-Ins and has been identified as an essential enzyme in Mycobacterium tuberculosis. We now report on the development of NTF1836 as a micromolar inhibitor of MshC. Using commercial libraries, we conducted preliminary structure-activity relationship (SAR) studies on NTF1836. Based on this data, NTF1836 and five structurally related compounds showed similar activity towards clinical strains of M. tuberculosis. A gram scale synthesis was developed to provide ample material for biological studies. Using this material, we determined that inhibition of M. tuberculosis growth by NTF1836 was accompanied by a fall in mycothiol and an increase in GlcN-Ins consistent with the targeting of MshC. We also determined that NTF1836 kills non-replicating M. tuberculosis in the carbon starvation model of latency.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, powder, γ-irradiated