Merck
Microporous Carbon Nanoparticles for Lithium-Sulfur Batteries.
Nanomaterials (Basel, Switzerland) (2020-10-16)
Hui-Ju Kang, Gazi A K M Rafiqul Bari, Tae-Gyu Lee, Tamal Tahsin Khan, Jae-Woo Park, Hyun Jin Hwang, Sung Yong Cho, Young-Si Jun
ABSTRACT
Rechargeable lithium-sulfur batteries (LSBs) are emerging as some of the most promising next-generation battery alternatives to state-of-the-art lithium-ion batteries (LIBs) due to their high gravimetric energy density, being inexpensive, and having an abundance of elemental sulfur (S8). However, one main, well-known drawback of LSBs is the so-called polysulfide shuttling, where the polysulfide dissolves into organic electrolytes from sulfur host materials. Numerous studies have shown the ability of porous carbon as a sulfur host material. Porous carbon can significantly impede polysulfide shuttling and mitigate the insulating passivation layers, such as Li2S, owing to its intrinsic high electrical conductivity. This work suggests a scalable and straightforward one-step synthesis method to prepare a unique interconnected microporous and mesoporous carbon framework via salt templating with a eutectic mixture of LiI and KI at 800 °C in an inert atmosphere. The synthesis step used environmentally friendly water as a washing solvent to remove salt from the carbon-salt mixture. When employed as a sulfur host material, the electrode exhibited an excellent capacity of 780 mAh g-1 at 500 mA g-1 and a sulfur loading mass of 2 mg cm-2 with a minor capacity loss of 0.36% per cycle for 100 cycles. This synthesis method of a unique porous carbon structure could provide a new avenue for the development of an electrode with a high retention capacity and high accommodated sulfur for electrochemical energy storage applications.
Sigma-Aldrich
Lithium iodide, AnhydroBeads, 99%
Sigma-Aldrich
Lithium nitrate, 99.99% trace metals basis