Merck
  • Lipophilic ion aromaticity is not important for permeability across lipid membranes.
Lipophilic ion aromaticity is not important for permeability across lipid membranes.
Biochimica et biophysica acta. Biomembranes (2020-10-02)
Tatyana I Rokitskaya, Elena A Kotova, Victor B Luzhkov, Roman S Kirsanov, Ekaterina V Aleksandrova, Galina A Korshunova, Vadim N Tashlitsky, Yuri N Antonenko
ABSTRACT
To clarify the contribution of charge delocalization in a lipophilic ion to the efficacy of its permeation through a lipid membrane, we compared the behavior of alkyl derivatives of triphenylphosphonium, tricyclohexylphosphonium and trihexylphosphonium both in natural and artificial membranes. Exploring accumulation of the lipophilic cations in response to inside-negative membrane potential generation in mitochondria by using an ion-selective electrode revealed similar mitochondrial uptake of butyltricyclohexylphosphonium (C4TCHP) and butyltriphenylphosphonium (C4TPP). Fluorescence correlation spectroscopy also demonstrated similar membrane potential-dependent accumulation of fluorescein derivatives of tricyclohexyldecylphosphonium and decyltriphenylphosphonium in mitochondria. The rate constant of lipophilic cation translocation across the bilayer lipid membrane (BLM), measured by the current relaxation method, moderately increased in the following sequence: trihexyltetradecylphosphonium ([P6,6,6,14]) < triphenyltetradecylphosphonium (C14TPP) < tricyclohexyldodecylphosphonium (C12TCHP). In line with these results, measurements of the BLM stationary conductance indicated that membrane permeability for C4TCHP is 2.5 times higher than that for C4TPP. Values of the difference in the free energy of ion solvation in water and octane calculated using the density functional theory and the polarizable continuum solvent model were similar for methyltriphenylphosphonium, tricyclohexylmethylphosphonium and trihexylmethylphosphonium. Our results prove that both cyclic and aromatic moieties are not necessary for lipophilic ions to effectively permeate through lipid membranes.
Sigma-Aldrich
Trihexyltetradecylphosphonium chloride, ≥95.0% (NMR)
Sigma-Aldrich
Dicyclohexylphenylphosphine, 95%