Merck
  • Activation-Induced Cytidine Deaminase Promotes Proliferation and Enhances Chemoresistance and Migration in B-cell Lymphoma.
Activation-Induced Cytidine Deaminase Promotes Proliferation and Enhances Chemoresistance and Migration in B-cell Lymphoma.
Anticancer research (2021-01-10)
Grant Godsmark, Leonardo Alves DE Souza Rios, Shaheen Mowla
ABSTRACT
Activation-induced cytidine deaminase (AID) is a DNA modifying enzyme which has an essential function in promoting antibody diversification. Its overexpression is strongly associated with B-cell derived malignancies including Burkitt lymphoma, where AID is required for the characteristic c-MYC/IGH translocation. This study aimed at defining AID's oncopathogenic role which is still poorly understood. We created over-expressing and knock-down cell culture models of AID, and used cellular assays to provide insight into its contribution to lymphomagenesis. We showed that AID expression is highly specific to, and abundantly expressed in B-cell-derived cancers and that ectopic overexpression of AID leads to rapid cell death. Using a knock-down model, we revealed that AID expression significantly impacts genomic stability, proliferation, migration and drug resistance. AID is an important driver of lymphoma, impacting multiple cellular events, and is potentially a strong candidate for targeted therapy in lymphoma.