Merck
  • Lactobacillus salivarius LI01 encapsulated in alginate-pectin microgels ameliorates D-galactosamine-induced acute liver injury in rats.
Lactobacillus salivarius LI01 encapsulated in alginate-pectin microgels ameliorates D-galactosamine-induced acute liver injury in rats.
Applied microbiology and biotechnology (2020-07-16)
Aoxiang Zhuge, Bo Li, Yin Yuan, Longxian Lv, Yating Li, Jingjing Wu, Liya Yang, Xiaoyuan Bian, Kaicen Wang, Qiangqiang Wang, Ren Yan, Xueling Zhu, Lanjuan Li
ABSTRACT
Acute liver failure is a clinical emergency associated with high mortality. Accumulating evidence indicates that gut microbiota participates in the progression of liver injury, and preventive therapies based on altering gut microbiota are of great interest. Previous studies demonstrated that Lactobacillus salivarius LI01 attenuates hepatic injury, though efficiency in curtailed in the harsh environment in the gastrointestinal tract. In this study, a system to encapsulate LI01 in alginate-pectin (AP) microgels was investigated. Encapsulation significantly enhances probiotic viability for long-term storage and heat treatment, and in simulated gastrointestinal fluids (SGF or SIF) and bile salt solutions. Acute liver injury was induced in Sprague-Dawley (SD) rats by D-galactosamine (D-GaIN) injection following pretreatment with probiotics. Liver and gut barrier function, cytokines, liver and gut histology, bacterial translocation, and gut microbiota were assessed. Administration of encapsulated LI01 more effectively upregulates hepatic anti-inflammatory cytokine IL-10 and TLR-3, restores expressions of gut barrier biomarkers Claudin-1 and MUC2 and attenuates destruction of mucosal ultrastructure compared with unencapsulated probiotics pretreatment. Pretreatment with AP-LI01 microgels altered the microbial community, decreasing the abundance of pathogenic taxa Ruminiclostridium, Dorea and Ruminococcaceae_UCG-004 and enriching beneficial taxa Ruminococcaceae_UCG-014, Eubacterium, and Prevotella_1 that produce short-chain fatty acids. These results suggest that AP encapsulation of LI01 boosts viability and attenuates liver injury by reducing inflammation and restoring intestinal barrier function. These beneficial effects are probably due to alternation of gut flora. These findings provide new insight into encapsulation technology and prevention of liver failure. KEY POINTS: • Alginate-pectin encapsulation enhances the viability of Lactobacillus salivarius LI01 under simulated commercial conditions and simulated gastrointestinal environment. • AP-LI01 microgel attenuates hepatic and intestinal inflammation and restores gut barrier function. • AP-LI01 microgel alters gut microbial community with increased SCFAs producers and decreased pathogenic microbes. • Beneficial improvements after administration of probiotics are highly associated with alternation of gut microbial community.
Sigma-Aldrich
D-(+)-Galactosamine hydrochloride, ≥99% (HPLC)