Merck
  • Impact of Magnesium Stearate Content: Modeling of Drug Degradation Using a Modified Arrhenius Equation.
Impact of Magnesium Stearate Content: Modeling of Drug Degradation Using a Modified Arrhenius Equation.
Chemical & pharmaceutical bulletin (2020-11-03)
Kousuke Tamura, Makoto Ono, Takefumi Kawabe, Etsuo Yonemochi
ABSTRACT
To accelerate drug development, the pharmaceutical industry is working to shorten and improve studies on stability. The Accelerated Stability Assessment Program (ASAP) incorporating the humidity-corrected Arrhenius equation as an accelerated methodology has been proposed for both drug substances and drug products. In this study, the effect of magnesium stearate (MgSt) content on the chemical stability of acetylsalicylic acid was evaluated as a model system of drug-excipient compatibility studies using ASAP. In the acetylsalicylic acid powder blends, temperature and humidity showed a first-order linear response to the natural logarithm of the reaction rate constant, and MgSt content also showed a first-order linear response. A polynomial model was built in which temperature, humidity, and MgSt content were independent each other. The fitting index of the model, the coefficient of determination, was 0.9567, which was a good fit. In the long-term stability study (25 °C/60% relative humidity, 6 months), there was good agreement in total between measured values and model-predicted values. Using this model, we inferred that the degradation rates were depended on MgSt content at the fixed temperature and humidity because the micro-environmental pH of the excipient was catalytically affected. Applying this model equation can significantly reduce the duration of formulation design and stability studies and save time and costs in drug development.
Product Description
Supelco
Methoxyacetyl fentanyl hydrochloride solution, 100 μg/mL in methanol (as free base), certified reference material, ampule of 0.5 mL, Cerilliant®