Merck
  • Detection of methicillin-resistant Staphylococcus aureus persistence in osteoblasts using imaging flow cytometry.
Detection of methicillin-resistant Staphylococcus aureus persistence in osteoblasts using imaging flow cytometry.
MicrobiologyOpen (2020-04-03)
Dafne Bongiorno, Nicolò Musso, Lorenzo Mattia Lazzaro, Gino Mongelli, Stefania Stefani, Floriana Campanile
ABSTRACT
Methicillin-resistant S. aureus has been reported as the main pathogen involved in chronic infections, osteomyelitis, and prosthetic joint infections. The host/pathogen interaction is dynamic and requires several changes to promote bacterial survival. Here, we focused on the internalization and persistence behavior of well-characterized Staphylococcus aureus invasive strains belonging to the main ST-MRSA-SCCmec clones. To overcome the limitations of the cell culture method, we comparatively analyzed the ability of internalization within human MG-63 osteoblasts with imaging flow cytometry (IFC). After evaluation by cell culture assay, the MRSA clones in the study were all able to readily internalize at 3h postinfection, the persistence of intracellular bacteria was evaluated at 24h both by routine cell culture and IFC assay, after vancomycin-BODIPY staining. A statistical difference of persistence was found in ST5-SCCmecII (26.59%), ST228-SCCmecI (20.25%), ST8-SCCmecIV (19.52%), ST239-SCCmecIII (47.82%), and ST22-SCCmecIVh (50.55%) showing the same ability to internalize as ATCC12598 (51%), the invasive isolate used as control strain for invasion and persistence assays. We demonstrated that the intracellular persistence process depends on the total number of infected cells. Comparing our data obtained by IFC with those of the cell culture assay, we obtained greater reproducibility rates and a number of intracellular bacteria, with the advantage of analyzing live host cells. Moreover, with some limitations related to the lack of whole-genome sequencing analysis, we validated the different proclivities to persist in the main Italian HA-MRSA invasive isolates and our results highlighted the heterogeneity of the different clones to persist during cell infection.
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein