Merck
  • Epigenomic Reprogramming toward Mesenchymal-Epithelial Transition in Ovarian-Cancer-Associated Mesenchymal Stem Cells Drives Metastasis.
Epigenomic Reprogramming toward Mesenchymal-Epithelial Transition in Ovarian-Cancer-Associated Mesenchymal Stem Cells Drives Metastasis.
Cell reports (2020-12-10)
Huihui Fan, Huda I Atiya, Yeh Wang, Thomas R Pisanic, Tza-Huei Wang, Ie-Ming Shih, Kelly K Foy, Leonard Frisbie, Ronald J Buckanovich, Alison A Chomiak, Rochelle L Tiedemann, Scott B Rothbart, Chelsea Chandler, Hui Shen, Lan G Coffman
ABSTRACT
A role for cancer cell epithelial-to-mesenchymal transition (EMT) in cancer is well established. Here, we show that, in addition to cancer cell EMT, ovarian cancer cell metastasis relies on an epigenomic mesenchymal-to-epithelial transition (MET) in host mesenchymal stem cells (MSCs). These reprogrammed MSCs, termed carcinoma-associated MSCs (CA-MSCs), acquire pro-tumorigenic functions and directly bind cancer cells to serve as a metastatic driver/chaperone. Cancer cells induce this epigenomic MET characterized by enhancer-enriched DNA hypermethylation, altered chromatin accessibility, and differential histone modifications. This phenomenon appears clinically relevant, as CA-MSC MET is highly correlated with patient survival. Mechanistically, mirroring MET observed in development, MET in CA-MSCs is mediated by WT1 and EZH2. Importantly, EZH2 inhibitors, which are clinically available, significantly inhibited CA-MSC-mediated metastasis in mouse models of ovarian cancer.
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Deoxyribonucleic acid from herring sperm, single-stranded for hybridization
Sigma-Aldrich
N,O-Bis(trimethylsilyl)carbamate, ≥98.0% (T)