- Role of growth differentiation factor 11 in development, physiology and disease.
Role of growth differentiation factor 11 in development, physiology and disease.
Oncotarget (2017-11-09)
Yonghui Zhang, Yong Wei, Dan Liu, Feng Liu, Xiaoshan Li, Lianhong Pan, Yi Pang, Dilong Chen
PMID29113418
ABSTRACT
Growth differentiation factor (GDF11) is a member of TGF-β/BMP superfamily that activates Smad and non-Smad signaling pathways and regulates expression of its target nuclear genes. Since its discovery in 1999, studies have shown the involvement of GDF11 in normal physiological processes, such as embryonic development and erythropoiesis, as well as in the pathophysiology of aging, cardiovascular disease, diabetes mellitus, and cancer. In addition, there are contradictory reports regarding the role of GDF11 in aging, cardiovascular disease, diabetes mellitus, osteogenesis, skeletal muscle development, and neurogenesis. In this review, we describe the GDF11 signaling pathway and its potential role in development, physiology and disease.