Merck
  • Chondroprotective effects of 4-methylumbelliferone and hyaluronan synthase-2 overexpression involve changes in chondrocyte energy metabolism.
Chondroprotective effects of 4-methylumbelliferone and hyaluronan synthase-2 overexpression involve changes in chondrocyte energy metabolism.
The Journal of biological chemistry (2019-10-18)
Kenya Terabe, Yoshifumi Ohashi, Saho Tsuchiya, Shinya Ishizuka, Cheryl B Knudson, Warren Knudson
ABSTRACT
Hyaluronan is a critical component of articular cartilage and partially helps retain aggrecan within the extracellular matrix of this tissue. During osteoarthritis, hyaluronan and aggrecan loss are an early sign of tissue damage. However, our recent attempts to mimic hyaluronan loss with the hyaluronan inhibitor 4-methylumbelliferone (4MU) did not exacerbate arthritis-like features of in vitro models of arthritis, but surprisingly, caused the reverse (i.e. provided potent chondroprotection). Moreover, the protective effects of 4MU did not depend on its role as a hyaluronan inhibitor. To understand the molecular mechanism in 4MU-mediated chondroprotection, we considered recent studies suggesting that shifts in intracellular UDP-hexose pools promote changes in metabolism. To determine whether such metabolic shifts are associated with the mechanism of 4MU-mediated pro-catabolic inhibition, using molecular and metabolomics approaches, we examined whether bovine and human chondrocytes exhibit changes in the contribution of glycolysis and mitochondrial respiration to ATP production rates as well as in other factors that respond to or might drive these changes. Overexpression of either HA synthase-2 or 4MU effectively reduced dependence on glycolysis in chondrocytes, especially enhancing glycolysis use by interleukin-1β (IL1β)-activated chondrocytes. The reduction in glycolysis secondarily enhanced mitochondrial respiration in chondrocytes, which, in turn, rescued phospho-AMP-activated protein kinase (AMPK) levels in the activated chondrocytes. Other glycolysis inhibitors, unrelated to hyaluronan biosynthesis, namely 2-deoxyglucose and dichloroacetate, caused metabolic changes in chondrocytes equivalent to those elicited by 4MU and similarly protected both chondrocytes and cartilage explants. These results suggest that fluxes in UDP-hexoses alter metabolic energy pathways in cartilage.
Product Description
Millipore
Pronase® Protease, Streptomyces griseus
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture