Merck
  • ZNF-281 as the Potential Diagnostic Marker of Oral Squamous Cell Carcinoma.
ZNF-281 as the Potential Diagnostic Marker of Oral Squamous Cell Carcinoma.
Cancers (2021-06-03)
Anna Starzyńska, Bartosz Kamil Sobocki, Aleksandra Sejda, Monika Sakowicz-Burkiewicz, Olga Szot, Barbara Alicja Jereczek-Fossa
ABSTRACT
ZNF-281 is a zinc finger factor which can lead to cancer progression and metastasis. Its up-regulation reported in many cancers was correlated with metastasis and worsened patients' prognosis. This is the first study describing ZNF-281 in the context of OSCC. Oral tissue samples drawn from 66 OSCC patients and 36 control patients were collected to determine protein (using immunochemistry and the semi-quantitative H-score method) and mRNA expression levels (using the RT-qPCR reaction). Our aim was to assess the ZNF-281 expression level in OSCC and the control group. Moreover, we determined the impact of ZNF-281 on survival parameters and the association of diversified clinical parameters with ZNF-281 expression. Clinical factors such as grade, AJCC stage and radiotherapy have an impact on the ZNF-281 H-score level, whereas AJCC stage and grade have an influence on ZNF-281 mRNA expression. Our survival analysis indicated that the impact on overall survival is not statistically significant, and the prognostic potential of ZNF-281 is rather limited. Our findings show that both levels of the ZNF-281 H-score and mRNA are decreased in OSCC in comparison to normal tissue. Moreover, we estimated that the H-score can differentiate normal tissue from OSCC with a sensitivity of 97% and specificity of 93.7%.