Merck
  • Multicentric carpotarsal osteolysis syndrome (MCTO) with generalized high bone turnover and high serum RANKL: Response to denosumab.
Multicentric carpotarsal osteolysis syndrome (MCTO) with generalized high bone turnover and high serum RANKL: Response to denosumab.
Bone reports (2021-01-29)
Ravit Regev, Etienne B Sochett, Yesmino Elia, Ronald M Laxer, Damien Noone, Kristi Whitney-Mahoney, Kornelia Filipowski, Amer Shamas, Reza Vali
ABSTRACT
MCTO is a rare disorder, caused by mutations in the MafB gene, a negative regulator of receptor activator of nuclear factor-кB ligand (RANKL). Manifestations include carpal and tarsal osteolysis and renal failure. Pathophysiology is poorly understood, and no effective treatment is available. In this case report we describe a patient with MCTO (MafB, mutation c.206C>T, p.Ser69Leu), diagnosed at the age of 5 years. At 7 years, skeletal survey showed diffuse osteopenia. BMD was mildly reduced, and bone turnover markers increased. He was treated with denosumab, a human monoclonal RANKL inhibitor for two years. Each injection was followed by a marked reduction in C-telopeptide (CTX). Following denosumab his BMD and bone symptoms improved and the osteolysis stabilized. At the age of 13 years, osteoporosis was diagnosed using high resolution peripheral quantitative computed tomography (HRpQCT) and serum RANKL was found to be markedly increased. This initial experience suggests that the associated osteoporosis may be ameliorated by denosumab, although further study will be needed to understand the appropriate dose, frequency, and the extent of efficacy. Monitoring of CTX and bone specific alkaline phosphatase will be especially useful in this regard. Further study in other MCTO patients is also needed to determine whether high bone turnover is specific to this mutation or more common than previously appreciated. We propose a model in which osteolysis in this condition is strongly associated with the systemic osteoporosis.