Merck
  • The differing pathophysiologies that underlie COVID-19-associated perniosis and thrombotic retiform purpura: a case series.
The differing pathophysiologies that underlie COVID-19-associated perniosis and thrombotic retiform purpura: a case series.
The British journal of dermatology (2020-08-12)
C M Magro, J J Mulvey, J Laurence, S Sanders, A N Crowson, M Grossman, J Harp, G Nuovo
ABSTRACT
There are two distinctive acral manifestations of COVID-19 embodying disparate clinical phenotypes. One is perniosis occurring in mildly symptomatic patients, typically children and young adults; the second is the thrombotic retiform purpura of critically ill adults with COVID-19. To compare the clinical and pathological profiles of these two different cutaneous manifestations of COVID-19. We compared the light microscopic, phenotypic, cytokine and SARS-CoV-2 protein and RNA profiles of COVID-19-associated perniosis with that of thrombotic retiform purpura in critical patients with COVID-19. Biopsies of COVID-19-associated perniosis exhibited vasocentric and eccrinotropic T-cell- and monocyte-derived CD11c+ , CD14+ and CD123+ dendritic cell infiltrates. Both COVID-associated and idiopathic perniosis showed striking expression of the type I interferon-inducible myxovirus resistance protein A (MXA), an established marker for type I interferon signalling in tissue. SARS-CoV-2 RNA, interleukin-6 and caspase 3 were minimally expressed and confined to mononuclear inflammatory cells. The biopsies from livedo/retiform purpura showed pauci-inflammatory vascular thrombosis without any MXA decoration. Blood vessels exhibited extensive complement deposition with endothelial cell localization of SARS-CoV-2 protein, interleukin-6 and caspase 3; SARS-CoV-2 RNA was not seen. COVID-19-associated perniosis represents a virally triggered exaggerated immune reaction with significant type I interferon signaling. This is important to SARS-CoV-2 eradication and has implications in regards to a more generalized highly inflammatory response. We hypothesize that in the thrombotic retiform purpura of critically ill patients with COVID-19, the vascular thrombosis in the skin and other organ systems is associated with a minimal interferon response. This allows excessive viral replication with release of viral proteins that localize to extrapulmonary endothelium and trigger extensive complement activation.