- Generation of an induced pluripotent stem cell line (TRNDi005-A) from a Mucopolysaccharidosis Type IVA (MPS IVA) patient carrying compound heterozygous p.R61W and p.WT405del mutations in the GALNS gene.
Generation of an induced pluripotent stem cell line (TRNDi005-A) from a Mucopolysaccharidosis Type IVA (MPS IVA) patient carrying compound heterozygous p.R61W and p.WT405del mutations in the GALNS gene.
Stem cell research (2019-02-24)
Rong Li, Amanda Baskfield, Jeanette Beers, Jizhong Zou, Chengyu Liu, Carlos J Alméciga-Díaz, Wei Zheng
PMID30797135
ABSTRACT
Mucopolysaccharidosis type IVA (MPS IVA) is a rare genetic disease caused by mutations in the GALNS gene and is inherited in an autosomal recessive manner. GALNS encodes N-acetylgalactosamine-6-sulfatase that breaks down certain complex carbohydrates known as glycosaminoglycans (GAGs). Deficiency in this enzyme causes accumulation of GAGs in lysosomes of body tissues. A human induced pluripotent stem cell (iPSC) line was generated from dermal fibroblasts of a MPS IVA patient that has compound heterozygous mutations (p.R61W and p.WT405del) in the GALNS gene. This iPSC line offers a useful resource to study the disease pathophysiology and a cell-based model for drug development.
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit