Merck
  • Discovery of ligands for a novel target, the human telomerase RNA, based on flexible-target virtual screening and NMR.
Discovery of ligands for a novel target, the human telomerase RNA, based on flexible-target virtual screening and NMR.
Journal of medicinal chemistry (2008-10-28)
Irene Gómez Pinto, Christophe Guilbert, Nikolai B Ulyanov, Jay Stearns, Thomas L James
ABSTRACT
The human ribonucleoprotein telomerase is a validated anticancer drug target, and hTR-P2b is a part of the human telomerase RNA (hTR) essential for its activity. Interesting ligands that bind hTR-P2b were identified by iteratively using a tandem structure-based approach: docking of potential ligands from small databases to hTR-P2b via the program MORDOR, which permits flexibility in both ligand and target, with subsequent NMR screening of high-ranking compounds. A high percentage of the compounds tested experimentally were found via NMR to bind to the U-rich region of hTR-P2b; most have MW < 500 Da and are from different compound classes, and several possess a charge of 0 or +1. Of the 48 ligands identified, 24 exhibit a decided preference to bind hTR-P2b RNA rather than A-site rRNA and 10 do not bind A-site rRNA at all. Binding affinity was measured by monitoring RNA imino proton resonances for some of the compounds that showed hTR binding preference.
Sigma-Aldrich
4,4′-Dihydroxybenzophenone, 99%