Merck
  • Induced pluripotent stem cell line (INSAi002-A) from a Fabry Disease patient hemizygote for the rare p.W287X mutation.
Induced pluripotent stem cell line (INSAi002-A) from a Fabry Disease patient hemizygote for the rare p.W287X mutation.
Stem cell research (2020-05-11)
Ana Joana Duarte, Diogo Ribeiro, Renato Santos, Luciana Moreira, José Bragança, Olga Amaral
ABSTRACT
Fabry Disease (FD) is a multisystemic X-linked disorder that belongs to the group of lysosomal storage disorders (LSDs). Causal mutations on alpha-galactosidase A (α-Gal A) commonly lead to abnormal protein and consequently to FD. Since it is an X-linked disease, males are primarily affected. This work describes the generation of induced Pluripotent Stem Cells (iPSCs) from skin fibroblasts from a FD patient, using non-integrative episomal vectors. Differentiation of iPSCs can be applied to generate a variety of cell types with high degree of genetic complexity that would otherwise be difficult to obtain.
Sigma-Aldrich
Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, clone 7F9.2, from mouse
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse