Merck
  • Efficient preparation of c-di-AMP at gram-scale using an immobilized Vibrio cholerae dinucleotide cyclase DncV.
Efficient preparation of c-di-AMP at gram-scale using an immobilized Vibrio cholerae dinucleotide cyclase DncV.
Enzyme and microbial technology (2020-12-31)
Qichao Sun, Yun Lv, Chenhui Zhang, Weifang Wu, Rui Zhang, Chunyuan Zhu, Yao-Yao Li, Huiqing Yuan, Jing Zhu, Deyu Zhu
ABSTRACT
Cyclic di-AMP is a bacterial nucleotide second messenger and evaluated as a potential vaccine adjuvant candidate. Here, we report a practical and economical enzymatic method for gram-scale preparation of c-di-AMP using an immobilized Vibrio cholerae dinucleotide cyclase DncV. The method mainly includes four steps: preparation of DncV-immobilized resin, enzymatic synthesis of c-di-AMP, purification using macroporous absorption resin SP207, and desiccation using rotary evaporation and lyophilization. Enzymatic synthesis is the most critical step, and almost all substrate ATP was converted to c-di-AMP under an optimum condition in which 300 mL of 300 mM NH4Ac/NH3 pH 9.5 buffer supplemented with 20 mM MnCl2, 10 mM ATP and 4 mL of DncV-immobilized resin containing ∼19 mg DncV were incubated at 30 °C overnight. After purification, up to 1 g of the diammonium salt of c-di-AMP with weight purity of ≥98% was obtained as white powder, which corresponds to an overall yield of ∼80% based on the ATP input into the reaction. The method is easily performed in laboratory to prepare c-di-AMP on a gram scale and could be used in industry on a large scale.
Sigma-Aldrich
c-di-AMP sodium salt, ≥98% (HPLC)