Merck
  • Contactin-1 Is Reduced in Cerebrospinal Fluid of Parkinson's Disease Patients and Is Present within Lewy Bodies.
Contactin-1 Is Reduced in Cerebrospinal Fluid of Parkinson's Disease Patients and Is Present within Lewy Bodies.
Biomolecules (2020-08-19)
Madhurima Chatterjee, Inger van Steenoven, Evelien Huisman, Linda Oosterveld, Henk Berendse, Wiesje M van der Flier, Marta Del Campo, Afina W Lemstra, Wilma D J van de Berg, Charlotte E Teunissen
ABSTRACT
Synaptic degeneration is an early phenomenon in Parkinson's disease (PD) pathogenesis. We aimed to investigate whether levels of synaptic proteins contactin-1 and contactin-2 in cerebrospinal fluid (CSF) of PD patients are reduced compared to dementia with Lewy bodies (DLB) patients and controls and to evaluate their relationship with α-synuclein aggregation. Contactin-1 and -2 were measured in CSF from PD patients (n = 58), DLB patients (n = 72) and age-matched controls (n = 90). Contactin concentration differences between diagnostic groups were assessed by general linear models adjusted for age and sex. Contactin immunoreactivity was characterized in postmortem substantia nigra, hippocampus and entorhinal cortex tissue of PD patients (n = 4) and controls (n = 4), and its relation to α-syn aggregation was evaluated using confocal laser scanning microscopy. Contactin-1 levels were lower in PD patients (42 (36-49) pg/mL) compared to controls (52 (44-58) pg/mL, p = 0.003) and DLB patients (56 (46-67) pg/mL, p = 0.001). Contactin-2 levels were similar across all diagnostic groups. Within the PD patient group, contactin-1 correlated with t-α-syn, tTau and pTau (r = 0.30-0.50, p < 0.05), whereas contactin-2 only correlated with t-α-syn (r = 0.34, p = 0.03). Contactin-1 and -2 were observed within nigral and cortical Lewy bodies and clustered within bulgy Lewy neurites in PD brains. A decrease in CSF contactin-1 may reflect synaptic degeneration underlying Lewy body pathology in PD.
Sigma-Aldrich
Mayer′s Hematoxylin Solution