Merck
  • Ketone Body Signaling Mediates Intestinal Stem Cell Homeostasis and Adaptation to Diet.
Ketone Body Signaling Mediates Intestinal Stem Cell Homeostasis and Adaptation to Diet.
Cell (2019-08-24)
Chia-Wei Cheng, Moshe Biton, Adam L Haber, Nuray Gunduz, George Eng, Liam T Gaynor, Surya Tripathi, Gizem Calibasi-Kocal, Steffen Rickelt, Vincent L Butty, Marta Moreno-Serrano, Ameena M Iqbal, Khristian E Bauer-Rowe, Shinya Imada, Mehmet Sefa Ulutas, Constantine Mylonas, Mark T Whary, Stuart S Levine, Yasemin Basbinar, Richard O Hynes, Mari Mino-Kenudson, Vikram Deshpande, Laurie A Boyer, James G Fox, Christopher Terranova, Kunal Rai, Helen Piwnica-Worms, Maria M Mihaylova, Aviv Regev, Ömer H Yilmaz
ABSTRACT
Little is known about how metabolites couple tissue-specific stem cell function with physiology. Here we show that, in the mammalian small intestine, the expression of Hmgcs2 (3-hydroxy-3-methylglutaryl-CoA synthetase 2), the gene encoding the rate-limiting enzyme in the production of ketone bodies, including beta-hydroxybutyrate (βOHB), distinguishes self-renewing Lgr5+ stem cells (ISCs) from differentiated cell types. Hmgcs2 loss depletes βOHB levels in Lgr5+ ISCs and skews their differentiation toward secretory cell fates, which can be rescued by exogenous βOHB and class I histone deacetylase (HDAC) inhibitor treatment. Mechanistically, βOHB acts by inhibiting HDACs to reinforce Notch signaling, instructing ISC self-renewal and lineage decisions. Notably, although a high-fat ketogenic diet elevates ISC function and post-injury regeneration through βOHB-mediated Notch signaling, a glucose-supplemented diet has the opposite effects. These findings reveal how control of βOHB-activated signaling in ISCs by diet helps to fine-tune stem cell adaptation in homeostasis and injury.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
(±)-Sodium 3-hydroxybutyrate, ≥99.0% (NT)
Pricing and availability is not currently available.