Merck
  • A Comparison of Solubility, Stability, and Bioavailability between Astilbin and Neoastilbin Isolated from Smilax glabra Rhizoma.
A Comparison of Solubility, Stability, and Bioavailability between Astilbin and Neoastilbin Isolated from Smilax glabra Rhizoma.
Molecules (Basel, Switzerland) (2020-10-21)
Dan Zheng, Yi-Ting Ruan, Zhong-Ping Yin, Qing-Feng Zhang
ABSTRACT
Astilbin and neoastilbin are two flavonoid stereoisomers. In the present study, their solubility, stability, and bioavailability were compared in a rat. The results revealed that the water solubility of astilbin and neoastilbin was 132.72 μg/mL and 217.16 μg/mL, respectively. The oil-water distribution coefficient (log P) of astilbin and neoastilbin in simulated gastric fluid (SGF) was 1.57 and 1.39, and in simulated intestinal fluid (SIF) was 1.09 and 0.98, respectively. In SIF, about 78.6% astilbin remained after 4 h of incubation at 37 °C, while this value was 88.3% for neoastilbin. Most of the degraded astilbin and neoastilbin were isomerized into their cis-trans-isomer, namely neoisoastilbin and isoastilbin, respectively, and the decomposed parts were rare. For bioavailability comparison in a rat, an HPLC method for trace amounts of astilbin and neoastilbin determination in plasma was developed, and the pretreatment of plasma was optimized. A pharmacokinetic study showed that the absolute bioavailability of astilbin and neoastilbin in a rat showed no significant difference with values of 0.30% and 0.28%, respectively.
Product Description
Astilbin, phyproof® Reference Substance