Merck
Small molecules that delay S phase suppress a zebrafish bmyb mutant.
Nature chemical biology (2005-12-24)
Howard M Stern, Ryan D Murphey, Jennifer L Shepard, James F Amatruda, Christian T Straub, Kathleen L Pfaff, Gerhard Weber, John A Tallarico, Randall W King, Leonard I Zon
ABSTRACT
Bmyb is a ubiquitously expressed transcription factor involved in cellular proliferation and cancer. Loss of bmyb function in the zebrafish mutant crash&burn (crb) results in decreased cyclin B1 expression, mitotic arrest and genome instability. These phenotypic observations in crb mutants could be attributed to the decreased expression of cyclin B1, a cell-cycle regulatory protein that is responsible for driving cell progression from G2 through mitosis. To identify small molecules that interact with the bmyb pathway, we developed an embryo-based suppressor screening strategy. In 16 weeks we screened a diverse approximately 16,000 compound library, and discovered one previously unknown compound, persynthamide (psy, 1), that suppressed bmyb-dependent mitotic defects. Psy-treated embryos showed an S-phase delay, and knockdown of the cell-cycle checkpoint regulator ataxia telangiectasia--and Rad-related kinase (ATR) abrogated the suppression of crb. The DNA synthesis inhibitors aphidicolin (2) and hydroxyurea (3) also suppressed crb. S-phase inhibition upregulated cyclin B1 mRNA, promoting the progression of cells through mitosis. Our study demonstrates that chemical suppressor screening in zebrafish can identify compounds with cell-cycle activity and can be used to identify pathways that interact with specific cell-cycle phenotypes.
Sigma-Aldrich
Hydroxyurea, 98%, powder