Merck
  • Potent estrogen receptor ligands based on bisphenols with a globular hydrophobic core.
Potent estrogen receptor ligands based on bisphenols with a globular hydrophobic core.
Journal of medicinal chemistry (2005-06-10)
Yasuyuki Endo, Tomohiro Yoshimi, Kiminori Ohta, Tomoharu Suzuki, Shigeru Ohta
ABSTRACT
Candidate estrogen receptor (ER) ligands with two phenolic residues on a three-dimensional hydrophobic core structure (carborane, bicyclo[2.2.2]octene, or adamantane) were synthesized and biologically evaluated. The biological properties of the ligands were markedly dependent on the nature of the hydrophobic core structure. Bis(4-hydroxyphenyl)-o-carborane (6) was a partial agonist/antagonist for ER. 1,2-Bis(4-hydroxyphenyl)bicyclo[2.2.2]octene (10) exhibited potent agonist activity for ER, even though the two phenolic groups are located similarly to those of 6.
Sigma-Aldrich
Tamoxifen, ≥99%